Methyl cellulose, 8000 cPs (C=2%, H₂O)

Supplier: Thermo Scientific Chemicals

Synonyms: Cellulose methyl ether

43483-22 43483-36
CAAA43483-22EA 102.38 CAD
CAAA43483-22 CAAA43483-36
Methyl cellulose, 8000 cPs (C=2%, H₂O)
Methyl cellulose

MDL: MFCD00081763 Notes: Viscosity 7000-10,000 cPs for a 2% aqueous solution at 20°C


CAS No.: 9004-67-5 Physical Form: Powder Odor: Odorless Merck Reference: 14,6040 MDL No.: MFCD00081763

Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS Number: 9004-67-5
Merck Index: 14,6040

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR