Barium carbonate, tech.

Supplier: Thermo Scientific Chemicals
Warning

43478-22 43478-36 43478-A3 43478-A9
CAAA43478-22EA 47.4 CAD
CAAA43478-22 CAAA43478-36 CAAA43478-A3 CAAA43478-A9
Barium carbonate, tech.
Barium carbonate

Used in the manufacture of glass, brick, clay products, barium chemicals, barium ferrites, and production of photographic papers


CAS No.: 513-77-9 Molecular Formula: BaCO3 Formula Weight: 197.35 Physical Form: Powder Density: 4.43 Odor: Odorless Boiling Point: dec. Solubility: Almost insoluble in water. Soluble in NH4Cl solution, dilute HCl, HNO3, acetic acid. Insoluble in most alkalies Merck Reference: 14,969 MDL No.: MFCD00003448


Used in the manufacture of glass, brick, clay products, barium chemicals, barium ferrites, and production of photographic papers

Formula: BaCO₃
MW: 197.34 g/mol
Boiling Pt: Dec.
Melting Pt: 1350
Density: 4.43
Storage Temperature: Ambient
MDL Number: MFCD00003448
CAS Number: 513-77-9
EINECS: 208-167-3
UN: 1564
ADR: 6.1 III
Merck Index: 14,969

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR