Potassium bromide, ultra dry ≥99.98% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
43448-14
43448-22
43448-36
CAAA43448-14EA
203.82
CAD
CAAA43448-14
CAAA43448-22
CAAA43448-36
Potassium bromide, ultra dry ≥99.98% (metals basis)
Potassium bromide
MDL: MFCD00011358
Fieser: 21,358
Purity: 99.98%
CAS No.: 7758-02-3
Molecular Formula: KBr
Formula Weight: 119.01
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 2.75
Refractive Index: 1.559
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 730°
Boiling Point: 1435°
Merck Reference: 14,7618
MDL No.: MFCD00011358
Formula:
KBr Boiling Pt: 1435 °C Melting Pt: 730 °C Density: 2.75 Storage Temperature: Ambient |
MDL Number:
MFCD00011358 CAS Number: 7758-02-3 EINECS: 231-830-3 Merck Index: 14,7618 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...