Sodium iodide, ultra dry ≥99.98% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Hydriodic acid sodium salt, Sodium monoiodide, Ioduril
43447-14
43447-22
43447-36
CAAA43447-14EA
243.32
CAD
CAAA43447-14
CAAA43447-22
CAAA43447-36
Sodium iodide, ultra dry ≥99.98% (metals basis)
Sodium iodide
MDL: MFCD00003532
Fieser: 1,1087 4,456 6,543 7,338 10,365 12,449 13,282 14,289 21,404
Purity: 99.98%
CAS No.: 7681-82-5
Molecular Formula: NaI
Formula Weight: 149.89
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 3.67
Refractive Index: 1.7745
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 651°
Boiling Point: 1300-1304°
Merck Reference: 14,8631
MDL No.: MFCD00003532
Formula:
NaI MW: 149.89 g/mol Boiling Pt: 1300…1304 °C Melting Pt: 651 °C Density: 3.67 Storage Temperature: Ambient |
MDL Number:
MFCD00003532 CAS Number: 7681-82-5 EINECS: 231-679-3 UN: 3077 ADR: 9,III Merck Index: 14,8631 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...