Polyethylene glycol 8,000

Supplier: Thermo Scientific Chemicals

43443-22 43443-36 43443-A3 43443-A9
CAAA43443-22EA 42.34 CAD
CAAA43443-22 CAAA43443-36 CAAA43443-A3 CAAA43443-A9
Polyethylene glycol 8,000
Polyethylene glycol 8,000

MDL: MFCD01779615 Notes: M.W. range 7000 - 9000


CAS No.: 25322-68-3 Molecular Formula: H(OCH2CH2)nOH Physical Form: Powder Appearance: White Odor: Odorless pH: 5-7 Melting Point: 55-63° Merck Reference: 14,7568 MDL No.: MFCD01779615

Formula: H(OCH₂CH₂)nOH
Boiling Pt: 250 °C (1013 hPa)
Melting Pt: 55…63 °C
Density: 1.125 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD01779615
CAS Number: 25322-68-3
EINECS: 500-038-2
Merck Index: 14,7568

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR