Aluminium ≥99.9995% (metals basis), wire, Puratronic®, annealed, Ø 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

43427-G5 43427-G9 43427-H4
CAAA43427-G5EA 101.75 CAD
CAAA43427-G5 CAAA43427-G9 CAAA43427-H4
Aluminium ≥99.9995% (metals basis), wire, Puratronic®, annealed, Ø 1.0 mm (0.04 in)
Aluminium

MDL: MFCD00134029


Purity: 99.9995% CAS No.: 7429-90-5 Physical Form: ≈2.1g/m, Temper: soft Odor: Odorless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR