Aluminium 99,9995 Metals basis, foil, Puratronic®, annealed, Thickness: 0.25 mm (0.01 in)
Supplier: Thermo Scientific Chemicals
Synonyms:
Aluminum
43424-FI
43424-GH
43424-HP
CAAA43424-FIEA
111.71
CAD
CAAA43424-FI
CAAA43424-GH
CAAA43424-HP
Aluminium 99,9995 Metals basis, foil, Puratronic®, annealed, Thickness: 0.25 mm (0.01 in)
Aluminium
MDL: MFCD00134029
Purity: 99.9995%
CAS No.: 7429-90-5
Physical Form: ≈1.7g/50x50mm, Temper: soft
Odor: Odorless
MDL No.: MFCD00134029
Formula:
Al MW: 26.98 g/mol Boiling Pt: 2327 Melting Pt: 660 Density: 2.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...