Indium ≥99.998% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Supplier: Thermo Scientific Chemicals
43414-14
43414-22
CAAA43414-14EA
383.94
CAD
CAAA43414-14
CAAA43414-22
Indium ≥99.998% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Indium
MDL: MFCD00134048
Purity: 99.998%
CAS No.: 7440-74-6
Formula Weight: 114.818
Physical Form: ≈1.5g/slug
Density: 7.310G/CC
Odor: Odorless
Boiling Point: 2072 deg C
MDL No.: MFCD00134048
Formula:
In Boiling Pt: 2072 °C Melting Pt: 156.6 °C Density: 7.310G/CC Storage Temperature: Ambient |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...