Ruthenium(IV) oxide hydrate ≥99.99% (metals basis) Ru 54-60%, Premion®
Supplier: Thermo Scientific Chemicals
|
Warning
|
43403-03
43403-06
43403-14
CAAA43403-03EA
141.09
CAD
CAAA43403-03
CAAA43403-06
CAAA43403-14
Ruthenium(IV) oxide hydrate ≥99.99% (metals basis) Ru 54-60%, Premion®
Ruthenium (IV) oxide hydrate
MDL: MFCD00149846
Purity: 99.99%
CAS No.: 32740-79-7
Molecular Formula: RuO2.xH2O
Formula Weight: 133.07(anhy)
Physical Form: Powder
Appearance: Black
Melting Point: 1200° subl.
UN No.: 1479
MDL No.: MFCD00149846
Formula:
O₂Ru·H₂O Melting Pt: 1200 °C (subl.) Density: 6.97 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149846 CAS Number: 32740-79-7 EINECS: 234-840-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...