Methyl levulinate 99+%
Supplier: Thermo Scientific Chemicals
Warning
|
43397-14
43397-22
CAAA43397-14EA
87.06
CAD
CAAA43397-14
CAAA43397-22
CAAA43397-36
Methyl levulinate 99+%
Methyl levulinate
MDL: MFCD00017499
Purity: 99+%
CAS No.: 624-45-3
Molecular Formula: CH3COCH2CH2CO2CH3
Formula Weight: 130.14
Flash Point: 72°(161.6°F)
Physical Form: Liquid
Density: 1.05
Odor: Pleasant
Boiling Point: 191°
MDL No.: MFCD00017499
Formula:
C₆H₁₀O₃ Boiling Pt: 191 °C Melting Pt: –24 °C Density: 1,05 Flash Pt: 72 °C (161.6 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00017499 CAS Number: 624-45-3 EINECS: 210-846-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...