Indium ≥99.99% (metals basis), rods, Ø 2.38 mm (0.094 in)

Supplier: Thermo Scientific Chemicals

43388-RQ 43388-Z8
CAAA43388-RQEA 159.58 CAD
CAAA43388-RQ CAAA43388-Z8
Indium ≥99.99% (metals basis), rods, Ø 2.38 mm (0.094 in)
Indium

MDL: MFCD00134048


Purity: 99.99% CAS No.: 7440-74-6 Physical Form: ≈0.33g/cm Odor: Odorless MDL No.: MFCD00134048

Formula: In
MW: 114.82 g/mol
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 156 °C
Density: 7.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR