2-Bromo-5-(trifluoromethyl)pyridine ≥96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
43381-03
43381-06
CAAA43381-03EA
107.28
CAD
CAAA43381-03
CAAA43381-06
2-Bromo-5-(trifluoromethyl)pyridine ≥96%
2-Bromo-5-(trifluoromethyl)pyridine
MDL: MFCD00153086
Purity: 96%
CAS No.: 50488-42-1
Molecular Formula: C6H3BrF3N
Formula Weight: 226.00
Physical Form: Crystalline
Melting Point: 45-47°
Boiling Point: 78-81°/30mm
MDL No.: MFCD00153086
Formula:
C₆H₃BrF₃N MW: 226 g/mol Boiling Pt: 78…81 °C (30 mmHg) Melting Pt: 45…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153086 CAS Number: 50488-42-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...