Strontium iodide, anhydrous ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

43374-06 43374-14 43374-22
CAAA43374-06EA 139.67 CAD
CAAA43374-06 CAAA43374-14 CAAA43374-22
Strontium iodide, anhydrous ≥99.99% (metals basis)
Strontium iodide

MDL: MFCD00049557


Purity: 99.99% CAS No.: 10476-86-5 Molecular Formula: SrI2 Formula Weight: 341.43 Physical Form: Powder Density: 4.550 Appearance: White to pale yellow Odor: Odorless Melting Point: 515° Merck Reference: 14,8844 MDL No.: MFCD00049557

Formula: I₂Sr
Melting Pt: 515 °C
Density: 4.550
Storage Temperature: Ambient
MDL Number: MFCD00049557
CAS Number: 10476-86-5
EINECS: 233-972-1
UN: 3260
ADR: 8,II
Merck Index: 14,8844

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR