Erbium triiodide, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals

43372-09 43372-18
CAAA43372-09EA 319.16 CAD
CAAA43372-09 CAAA43372-18
Erbium triiodide, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®
Erbium triiodide

MDL: MFCD00016075


Purity: 99.99% CAS No.: 13813-42-8 Molecular Formula: ErI3 Formula Weight: 547.97 Physical Form: Powder Sensitivity: Hygroscopic Boiling Point: 1280° MDL No.: MFCD00016075

Formula: ErI₃
MW: 547.97 g/mol
Boiling Pt: 1280 °C
Melting Pt: 1020 °C
Density: ∼5.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016075
CAS Number: 13813-42-8
EINECS: 237-471-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR