Lithium wolframate ≥99%
Supplier: Thermo Scientific Chemicals
43366-18
43366-30
CAAA43366-18EA
164.32
CAD
CAAA43366-18
CAAA43366-30
Lithium wolframate ≥99%
Lithium wolframate
MDL: MFCD00016185
Purity: 99% min
CAS No.: 13568-45-1
Molecular Formula: Li2WO4
Formula Weight: 261.73
Physical Form: Powder
Density: 3.71
Melting Point: 742°
MDL No.: MFCD00016185
Formula:
Li₂O₄W Melting Pt: 742 °C Density: 3.71 Storage Temperature: Ambient |
MDL Number:
MFCD00016185 CAS Number: 13568-45-1 EINECS: 236-978-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...