Aluminium ≥99.9995% (metals basis), slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

43357-06 43357-14 43357-22
CAAA43357-06EA 31.6 CAD
CAAA43357-06 CAAA43357-14 CAAA43357-22
Aluminium ≥99.9995% (metals basis), slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Aluminium

MDL: MFCD00134029


Purity: 99.9995% CAS No.: 7429-90-5 Physical Form: ≈0.54g/slug Odor: Odorless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR