Palladium ≥99.95% (metals basis), granules

Supplier: Thermo Scientific Chemicals

43353-03 43353-06 43353-14
CAAA43353-03EA 522.98 CAD
CAAA43353-03 CAAA43353-06 CAAA43353-14
Palladium ≥99.95% (metals basis), granules
Palladium

MDL: MFCD00011167


Purity: 99.95% CAS No.: 7440-05-3 Odor: Odorless MDL No.: MFCD00011167

Formula: Pd
Boiling Pt: 3167 °C (1013 hPa)
Melting Pt: 1555 °C
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
Merck Index: 12,07121

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR