Tin ≥99.98% (metals basis), rods, Ø 12.7 mm (0.5 in)

Supplier: Thermo Scientific Chemicals

43341-BU 43341-BY
CAAA43341-BUEA 184.86 CAD
CAAA43341-BU CAAA43341-BY
Tin ≥99.98% (metals basis), rods, Ø 12.7 mm (0.5 in)
Tin

MDL: MFCD00133862


Purity: 99.98% CAS No.: 7440-31-5 Molecular Formula: Sn Formula Weight: 118.710 Physical Form: ≈9.23g/cm Density: 7.310 MDL No.: MFCD00133862

Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.310
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR