Aluminium ≥99.999% (metals basis), wire, Puratronic®, annealed, Ø 0.81 mm (0.032 in)

Supplier: Thermo Scientific Chemicals

43336-G6 43336-H2 43336-H7
CAAA43336-G6EA 99.7 CAD
CAAA43336-G6 CAAA43336-H2 CAAA43336-H7
Aluminium ≥99.999% (metals basis), wire, Puratronic®, annealed, Ø 0.81 mm (0.032 in)
Aluminium

MDL: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR