Silicon carbide ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

43332-36 43332-A3
CAAA43332-36EA 211.72 CAD
CAAA43332-36 CAAA43332-A3
Silicon carbide ≥99% (metals basis)
Silicon carbide

MDL: MFCD00049531


Purity: 99% CAS No.: 409-21-2 Molecular Formula: CSi Formula Weight: 40.10 Physical Form: -325 Mesh Powder, Black Density: 3.16 Appearance: Black Odor: Odorless Melting Point: 2700° Merck Reference: 14,8492 MDL No.: MFCD00049531

Formula: SiC
Melting Pt: 2700 °C
Density: 3.16
Storage Temperature: Ambient
MDL Number: MFCD00049531
CAS Number: 409-21-2
EINECS: 206-991-8
Merck Index: 14,8492
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bd532b391b9c28edcb8adac5fe5ed958ccaa487&vid=d39ce1cfd6c1a5ca44c7d11b656758cbfaed190d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR