Platinum iridium alloy (80:20 w%) ≥99.9% (metals basis excluding precious metals), wire, hard, Ø 0.127 mm (0.005 in)
Supplier: Thermo Scientific Chemicals
43330-BS
43330-BW
CAAA43330-BSEA
145.36
CAD
CAAA43330-BS
CAAA43330-BW
Platinum iridium alloy (80:20 w%) ≥99.9% (metals basis excluding precious metals), wire, hard, Ø 0.127 mm (0.005 in)
Platinum Iridium alloy (80:20 w%)
MDL: MFCD02091683
Purity: 99.9%
Molecular Formula: Pt:Ir; 80:20 wt%
Physical Form: ≈0.03g/10cm, Temper: hard
Odor: Odorless
MDL No.: MFCD02091683
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...