L(+)-Aspartic acid 99% low metals

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-(+)-Aminosuccinic acid (S)-Aminobutanedioic acid (S)-Aminosuccinic acid H-Asp-OH L-Asparaginic acid L(+)-Aspartic acid, (S)-(+)-Aspartic acid, L(+)-2-Amino-1,4-butanedioic acid, (S)-(+)-2-Amino-1,4-butanedioic acid, L-Aspartic acid, H-Asp-OH, L-α-Aminosuccinic acid, L-αlpha-Aminosuccinic acid, (S)-2-Aminosuccinic acid, Asp, D

43317-22 43317-36
CAAA43317-22EA 60.36 CAD
CAAA43317-22 CAAA43317-36
L(+)-Aspartic acid 99% low metals
L(+)-Aspartic acid

MDL: MFCD00002616 Beilstein Registry No.: 1723530 Notes: Specific rotation (4%, 6NHCl) +24.5°C to 26.0°C, As less than 10ppm, Feless than 20ppm


Purity: 99% CAS No.: 56-84-8 Molecular Formula: C4H7NO4 Formula Weight: 133.10 Physical Form: Powder Density: 1.66 Melting Point: 300° dec. Beilstein Reference: 1723530 Merck Reference: 14,840 MDL No.: MFCD00002616

Formula: C₄H₇NO₄
MW: 133.1 g/mol
Boiling Pt: 311
Melting Pt: >300 °C (dec.)
Density: 1.66
Storage Temperature: Ambient
MDL Number: MFCD00002616
CAS Number: 56-84-8
EINECS: 200-291-6
Merck Index: 14,840

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR