Tin ≥99.9% (metals basis), foil, Thickness: 0.18 mm (0.007 in)

Supplier: Thermo Scientific Chemicals

43308-FI 43308-GH 43308-HP
CAAA43308-FIEA 96.38 CAD
CAAA43308-FI CAAA43308-GH CAAA43308-HP
Tin ≥99.9% (metals basis), foil, Thickness: 0.18 mm (0.007 in)
Tin

MDL: MFCD00133862


Purity: 99.9% CAS No.: 7440-31-5 Physical Form: ≈3.3g/50x50mm Odor: Odorless MDL No.: MFCD00133862

Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270
Melting Pt: 232
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR