Magnesium ≥99.95% (metals basis), slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

43299-09 43299-18 43299-30
CAAA43299-09EA 102.54 CAD
CAAA43299-09 CAAA43299-18 CAAA43299-30
Magnesium ≥99.95% (metals basis), slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Magnesium

MDL: MFCD00085308


Purity: 99.95% CAS No.: 7439-95-4 Physical Form: ≈0.7g/slug Odor: Odorless MDL No.: MFCD00085308

Formula: Mg
Boiling Pt: 1107 °C (1013 hPa)
Melting Pt: 651 °C
Density: 1.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III
Merck Index: 13,05675

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR