Magnesium 99,95 Metals basis, slug, annealed, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Supplier: Thermo Scientific Chemicals
43297-09
43297-18
CAAA43297-09EA
216.46
CAD
CAAA43297-09
CAAA43297-18
Magnesium 99,95 Metals basis, slug, annealed, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Magnesium
MDL: MFCD00085308
Purity: 99.95%
CAS No.: 7439-95-4
Physical Form: ≈0.09g/slug
Odor: Odorless
MDL No.: MFCD00085308
Formula:
Mg MW: 24.3 g/mol Boiling Pt: 1107 Melting Pt: 651 Density: 1.74 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00085308 CAS Number: 7439-95-4 EINECS: 231-104-6 UN: 1869 ADR: 4.1 III Merck Index: 13,05675 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...