Magnesium ≥99.95% (metals basis), slug, unannealed, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

43296-09 43296-18
CAAA43296-09EA 246.48 CAD
CAAA43296-09 CAAA43296-18
Magnesium ≥99.95% (metals basis), slug, unannealed, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Magnesium

MDL: MFCD00085308


Purity: 99.95% CAS No.: 7439-95-4 Physical Form: ≈0.04g/slug Odor: Odorless MDL No.: MFCD00085308

Formula: Mg
Boiling Pt: 1107 °C (1013 hPa)
Melting Pt: 651 °C
Density: 1.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III
Merck Index: 13,05675

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR