Rhenium pentachloride ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Rhenium(V) chloride
43294-04
43294-09
CAAA43294-04EA
669.92
CAD
CAAA43294-04
CAAA43294-09
Rhenium pentachloride ≥99.9% (metals basis)
Rhenium pentachloride
MDL: MFCD00011197
Purity: 99.9%
CAS No.: 13596-35-5
Molecular Formula: ReCl5
Formula Weight: 363.47
Physical Form: -50 Mesh Powder
Sensitivity: Moisture Sensitive
MDL No.: MFCD00011197
Formula:
Cl₅Re MW: 363.47 g/mol Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00011197 CAS Number: 13596-35-5 EINECS: 237-042-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...