Platinum iridium alloy (85:15 w%), wire, annealed, Ø 0.25 mm (0.01 in)
Supplier: Thermo Scientific Chemicals
43275-BU
43275-BY
CAAA43275-BUEA
342.86
CAD
CAAA43275-BU
CAAA43275-BY
Platinum iridium alloy (85:15 w%), wire, annealed, Ø 0.25 mm (0.01 in)
Platinum Iridium alloy (85:15 w%)
MDL: MFCD02091683
Molecular Formula: Pt:Ir; 85:15 wt%
Physical Form: ≈0.27g/25cm, Temper: soft
Odor: Odorless
MDL No.: MFCD02091683
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...