Copper nickel alloy (55:45 w%), wire, Ø 0.5 mm (0.02 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
43272-G9
43272-H4
43272-H9
CAAA43272-G9EA
64.94
CAD
CAAA43272-G9
CAAA43272-H4
CAAA43272-H9
Copper nickel alloy (55:45 w%), wire, Ø 0.5 mm (0.02 in)
Copper Nickel alloy (55:45 w%)
MDL: MFCD00801098
Molecular Formula: Cu:Ni 55:45 wt%
Physical Form: ≈1.75g/m
MDL No.: MFCD00801098
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...