Copper nickel alloy (55:45 w%), wire, Ø 0.25 mm (0.01 in)
Supplier: Thermo Scientific Chemicals
43271-H4
43271-H9
CAAA43271-H4EA
85.64
CAD
CAAA43271-H4
CAAA43271-H9
Copper nickel alloy (55:45 w%), wire, Ø 0.25 mm (0.01 in)
Copper Nickel alloy (55:45 w%)
MDL: MFCD00801098
Molecular Formula: Cu:Ni 55:45 wt%
Physical Form: ≈0.44g/m
MDL No.: MFCD00801098
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...