Copper nickel alloy (55:45 w%), wire, Ø 0.13 mm (0.005 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
43270-H4
43270-H9
CAAA43270-H4EA
56.72
CAD
CAAA43270-H4
CAAA43270-H9
Copper nickel alloy (55:45 w%), wire, Ø 0.13 mm (0.005 in)
Copper Nickel alloy (55:45 w%)
MDL: MFCD00801098
Molecular Formula: Cu:Ni 55:45 wt%
Physical Form: ≈0.11g/m
Odor: Odorless
MDL No.: MFCD00801098
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...