Copper nickel alloy (55:45 w%), wire, Ø 0.075 mm (0.003 in)
Supplier: Thermo Scientific Chemicals
43269-H4
43269-H9
CAAA43269-H4EA
56.56
CAD
CAAA43269-H4
CAAA43269-H9
Copper nickel alloy (55:45 w%), wire, Ø 0.075 mm (0.003 in)
Copper Nickel alloy (55:45 w%)
Resistance alloy, negative lead for type J thermocouple
Molecular Formula: Cu:Ni 55:45 wt%
Physical Form: ≈0.04g/m
MDL No.: MFCD00801098
Resistance alloy, negative lead for type J thermocouple
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...