Copper ≥99.99% (metals basis), wire, Ø 0.015 mm (0.0006 in)

Supplier: Thermo Scientific Chemicals

43238-G6 43238-H2
CAAA43238-G6EA 598.82 CAD
CAAA43238-G6 CAAA43238-H2
Copper ≥99.99% (metals basis), wire, Ø 0.015 mm (0.0006 in)
Copper

MDL: MFCD00010965

Formula: Cu
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR