Tin ≥99.8% (metals basis), foil, Thickness: 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals

43234-FI 43234-GH 43234-HP
CAAA43234-FIEA 63.04 CAD
CAAA43234-FI CAAA43234-GH CAAA43234-HP
Tin ≥99.8% (metals basis), foil, Thickness: 0.5 mm (0.02 in)
Tin

MDL: MFCD00133862


Purity: 99.8% CAS No.: 7440-31-5 Physical Form: ≈9.1g/50x50mm Odor: Odorless MDL No.: MFCD00133862

Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR