Tin ≥99.8% (metals basis), foil, Thickness: 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

43233-FI 43233-GH 43233-HP
CAAA43233-FIEA 53.09 CAD
CAAA43233-FI CAAA43233-GH CAAA43233-HP
Tin ≥99.8% (metals basis), foil, Thickness: 0.25 mm (0.01 in)
Tin

MDL: MFCD00133862


Purity: 99.8% CAS No.: 7440-31-5 Physical Form: ≈4.6g/50x50mm Odor: Odorless MDL No.: MFCD00133862

Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270
Melting Pt: 232
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR