Magnesium oxide ≥95%

Supplier: Thermo Scientific Chemicals

Synonyms: Magnesia usta

43195-22 43195-36 43195-A3
CAAA43195-22EA 115.81 CAD
CAAA43195-22 CAAA43195-36 CAAA43195-A3
Magnesium oxide ≥95%
Magnesium oxide

For plasma spraying, Refractory applications, electrical insulation, pharmaceuticals, cosmetics, paper manufacture, adsorption and catalysis, and in semiconductors, reflector in optical instruments


Purity: 95% min CAS No.: 1309-48-4 Molecular Formula: MgO Formula Weight: 40.30 Physical Form: -140+325 Mesh Powder Density: 3.58 Refractive Index: 1.736 Odor: Odorless Melting Point: 2852° Boiling Point: 3600° Solubility: Slightly soluble in water, (solubility increased by CO2). Soluble in acids and ammonium salt solution. Insoluble in alcohol Merck Reference: 14,5677 MDL No.: MFCD00011109


For plasma spraying, Refractory applications, electrical insulation, pharmaceuticals, cosmetics, paper manufacture, adsorption and catalysis, and in semiconductors, reflector in optical instruments

Formula: MgO
MW: 40.3 g/mol
Boiling Pt: 3600
Melting Pt: 2800
Density: 3.58
Storage Temperature: Ambient
MDL Number: MFCD00011109
CAS Number: 1309-48-4
EINECS: 215-171-9
Merck Index: 14,5677

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR