1,4-Dioxane, anhydrous ≥99%, chemSeal™ for HPLC

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide

Packaged under argon in resealable ChemSeal™ bottles.

43167-AK 43167-K2
CAAA43167-AKEA 90.22 CAD
CAAA43167-AK CAAA43167-K2
1,4-Dioxane, anhydrous ≥99%, chemSeal™ for HPLC
1,4-Dioxane

MDL: MFCD00006571 Beilstein Registry No.: 102551 Notes: Specifications: Same HPLC specifications as 22913 Fieser: 1,333


Purity: 99% min CAS No.: 123-91-1 Molecular Formula: C4H8O2 Formula Weight: 88.11 Flash Point: 12°(54°F) Physical Form: Liquid Density: 1.034 Refractive Index: 1.4220 Sensitivity: Hygroscopic Odor: Ether-like Melting Point: 11.8° Boiling Point: 100-102° Beilstein Reference: 102551 Merck Reference: 14,3300 UN No.: 1165 MDL No.: MFCD00006571

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 101.1
Melting Pt: 11.8
Density: 1.034
Flash Pt: 12 °C (54 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006571
CAS Number: 123-91-1
EINECS: 204-661-8
UN: 1165
ADR: 3 II
Merck Index: 14,3300
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR