4-Octyne 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
43157-06
43157-14
CAAA43157-06EA
63.83
CAD
CAAA43157-06
CAAA43157-14
4-Octyne 99%
4-Octyne
MDL: MFCD00009471
Purity: 99%
CAS No.: 1942-45-6
Molecular Formula: CH3CH2CH2C≡CCH2CH2CH3
Formula Weight: 110.20
Flash Point: 20°(68°F)
Physical Form: Liquid
Density: 0.7510
Refractive Index: 1.4236
Odor: Petroleum-like
Boiling Point: 131-132°
UN No.: 3295
MDL No.: MFCD00009471
Formula:
C₈H₁₄ Boiling Pt: 131…132 °C Melting Pt: –103 °C Density: 0.7510 Flash Pt: 20 °C (68 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009471 CAS Number: 1942-45-6 EINECS: 217-730-2 UN: 3295 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...