cis-4-Octene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (Z)-4-Octene

43156-04 43156-09 43156-18
CAAA43156-04EA 77.89 CAD
CAAA43156-04 CAAA43156-09 CAAA43156-18
cis-4-Octene 97%
cis-4-Octene

MDL: MFCD00067484


Purity: 97% CAS No.: 7642-15-1 Molecular Formula: C8H16 Formula Weight: 112.22 Physical Form: Liquid UN No.: 3295 MDL No.: MFCD00067484

Formula: C₈H₁₆
MW: 112.22 g/mol
Boiling Pt: 123 °C (1013 hPa)
Melting Pt: –118.7 °C
Density: 0.7212 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00067484
CAS Number: 7642-15-1
EINECS: 231-578-4
UN: 3295
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR