trans-4-Octene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(E)-4-Octene
43155-04
43155-09
43155-18
CAAA43155-04EA
62.25
CAD
CAAA43155-04
CAAA43155-09
CAAA43155-18
trans-4-Octene 97%
trans-4-Octene
MDL: MFCD00009476
Beilstein Registry No.: 1719104
Purity: 97%
CAS No.: 14850-23-8
Molecular Formula: C8H16
Formula Weight: 112.22
Flash Point: 21°(70°F)
Physical Form: Liquid
Density: 0.715
Refractive Index: 1.4120
Odor: Characteristic
Boiling Point: 122-123°
Beilstein Reference: 1719104
UN No.: 3295
MDL No.: MFCD00009476
Formula:
C₈H₁₆ Boiling Pt: 122…123 °C Melting Pt: –93.8 °C Density: 0,715 Flash Pt: 21 °C (70 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009476 CAS Number: 14850-23-8 EINECS: 238-913-3 UN: 3295 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...