Methyl cellulose, 400 cP (C=2%, H₂O)
Supplier: Thermo Scientific Chemicals
Synonyms:
Cellulose methyl ether
43147-22
43147-36
43147-A4
CAAA43147-22EA
68.41
CAD
CAAA43147-22
CAAA43147-36
CAAA43147-A4
Methyl cellulose, 400 cP (C=2%, H₂O)
Methyl cellulose
MDL: MFCD00081763
Notes: Viscosity: 300-560 cPs for a 2% aqueous solution at 20°C
CAS No.: 9004-67-5
Physical Form: Powder
Odor: Odorless
Merck Reference: 14,6040
MDL No.: MFCD00081763
Storage Temperature:
Ambient |
MDL Number:
MFCD00081763 CAS Number: 9004-67-5 Merck Index: 14,6040 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...