Methyl cellulose, 1,600 cPs (C=2%, H₂O)

Supplier: Thermo Scientific Chemicals

Synonyms: Cellulose methyl ether

43146-22 43146-36
CAAA43146-22EA 70.63 CAD
CAAA43146-22 CAAA43146-36 CAAA43146-A4
Methyl cellulose, 1,600 cPs (C=2%, H₂O)
Methyl cellulose

MDL: MFCD00081763 Notes: Viscosity: 1125-2100 cPs for a 2% aqueous solution at 20°C


CAS No.: 9004-67-5 Physical Form: Powder Odor: Odorless Merck Reference: 14,6040 MDL No.: MFCD00081763

Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS Number: 9004-67-5
Merck Index: 14,6040

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR