Aluminium nitrate nonahydrate ≥99.999% (metals basis), Puratronic® low in mercury
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Aluminum nitrate nonahydrate
43145-14
43145-22
43145-36
CAAA43145-14EA
208.56
CAD
CAAA43145-14
CAAA43145-22
CAAA43145-36
Aluminium nitrate nonahydrate ≥99.999% (metals basis), Puratronic® low in mercury
Aluminium nitrate nonahydrate
MDL: MFCD00003419
Purity: 99.999%
CAS No.: 7784-27-2
Molecular Formula: Al(NO3)3.9H2O
Formula Weight: 375.13 (212.99anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Merck Reference: 14,352
UN No.: 1438
MDL No.: MFCD00003419
Formula:
Al(NO₃)₃·9H₂O Melting Pt: 73.5 °C Density: 1.72 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149132 CAS Number: 7784-27-2 EINECS: 236-751-8 UN: 1438 ADR: 5.1,III Merck Index: 14,352 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...