5-Sulfosalicylic acid dihydrate 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Hydroxy-5-sulfobenzoic acid dihydrate, 2-Hydroxy-5-sulphobenzoic acid dihydrate
Reagent for precipitation of proteins
Purity: 99+%
CAS No.: 5965-83-3
Molecular Formula: C7H6O6S.2H2O
Formula Weight: 254.22 (218.19anhy)
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: White
Odor: Pungent
Melting Point: 103-112°
Beilstein Reference: 650741
Merck Reference: 14,8964
UN No.: 2585
MDL No.: MFCD00007508
Reagent for precipitation of proteins
Formula:
HO₃SC₆H₃-2-(OH)CO₂H·2H₂O MW: 254.22 g/mol Melting Pt: 103…112 °C Density: 1.695 g/cm³ (20 °C) Flash Pt: 150 Storage Temperature: Ambient |
MDL Number:
MFCD00149540 CAS Number: 5965-83-3 EINECS: 202-555-6 UN: 2585 ADR: 8 III Merck Index: 14,8964 |
|
Specification Test Results
Insoluble matter | 0.02% |
Residue after ignition | 0.1 |
Cl | 0.001% |
HOC†H3COOH | 0.04% |
SO3 | 0.02% |
PB | 0.002% |
Fe | 0.001% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...