2,3-Dimethylbutane 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
43142-K2
43142-AK
CAAAA43142-K2EA
1074.4
CAD
CAAAA43142-K2
CAAA43142-AK
2,3-Dimethylbutane 98+%
2,3-Dimethylbutane
MDL: MFCD00008925
Beilstein Registry No.: 1730737
Purity: 98+%
CAS No.: 79-29-8
Molecular Formula: C6H14
Formula Weight: 86.18
Flash Point: -29°(-20°F)
Physical Form: Liquid
Density: 0.662
Refractive Index: 1.375
Appearance: Colorless
Odor: Mild
Melting Point: -129°
Boiling Point: 57-59°
Beilstein Reference: 1730737
UN No.: 2457
MDL No.: MFCD00008925
Formula:
C₆H₁₄ Boiling Pt: 57…59 °C Melting Pt: –129 °C Density: 0,662 Flash Pt: –29 °C (–20 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008925 CAS Number: 79-29-8 EINECS: 201-193-6 UN: 2457 ADR: 3,II |
Specification Test Results
Form | Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...