Lanthanum(III) hydroxide ≥99.95% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

43123-18 43123-30
CAAA43123-18EA 108.7 CAD
CAAA43123-18 CAAA43123-30
Lanthanum(III) hydroxide ≥99.95% (REO, rare earth oxide basis)
Lanthanum (III) hydroxide

MDL: MFCD00016160


Purity: 99.95% CAS No.: 14507-19-8 Molecular Formula: La(OH)3 Formula Weight: 189.93 Physical Form: Powder Appearance: White Odor: Odorless Melting Point: dec. MDL No.: MFCD00016160

Formula: H₃LaO₃
MW: 189.93 g/mol
Melting Pt: Dec.
Density: 4,28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016160
CAS Number: 14507-19-8
EINECS: 238-510-2
UN: 3262
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR