Aluminium oxide, aerosol refractory brushable paint

Supplier: Thermo Scientific Chemicals
Danger

43119-30 43119-A1
CAAA43119-30EA 161.16 CAD
CAAA43119-30 CAAA43119-A1
Aluminium oxide, aerosol refractory brushable paint
Aluminium oxide

Provides protective high temperature Al2O3 coating on ceramic, graphite, or metal


CAS No.: 1344-28-1 Molecular Formula: Al2O3 Flash Point: -4°(25°F) Physical Form: Paste Appearance: White UN No.: 1263 MDL No.: MFCD00003424


Provides protective high temperature Al2O3 coating on ceramic, graphite, or metal

Formula: Al₂O₃
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3.94 g/cm³ (20 °C)
Flash Pt: –4 °C (25 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
UN: 1263
ADR: 3,II
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR