Charcoal activated -4+8 mesh

Supplier: Thermo Scientific Chemicals

43118-22 43118-36 43118-A3
CAAA43118-22EA 47.24 CAD
CAAA43118-22 CAAA43118-36 CAAA43118-A3
Charcoal activated -4+8 mesh
Charcoal activated

MDL: MFCD00133992


MDL No.: MFCD00133992

Formula: C
Melting Pt: 3550 °C
Density: 2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR