Silicon dioxide 30% in water, colloidal dispersion

Supplier: Thermo Scientific Chemicals

43111-A1 43111-A7
CAAA43111-A1EA 52.14 CAD
CAAA43111-A1 CAAA43111-A7 CAAA43111-C4
Silicon dioxide 30% in water, colloidal dispersion
Silicium dioxide

As a binder in ceramic investment shell coating, with refractory powders for yielding superior face coats


CAS No.: 7631-86-9 Molecular Formula: O2Si Formula Weight: 60.09 Physical Form: 0.01 Micron Particles, in Liquid, S.A. 305m2/g Density: 1.20 Appearance: Colorless MDL No.: MFCD00011232


As a binder in ceramic investment shell coating, with refractory powders for yielding superior face coats

Formula: SiO₂
Density: 1.20
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9
EINECS: 231-545-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR