Silicon dioxide 50% in water, colloidal dispersion

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide

43109-A1 43109-A7
CAAA43109-A1EA 48.35 CAD
CAAA43109-A1 CAAA43109-A7
Silicon dioxide 50% in water, colloidal dispersion
Silicium dioxide

MDL: MFCD00011232 Notes: Particle charge (-), pH 10.0, viscosity 50 cps


CAS No.: 7631-86-9 Molecular Formula: SiO2 Formula Weight: 60.09 Physical Form: 0.02 Micron Particles, in Liquid, S.A. 180m2/g Density: 1.40 Odor: Odorless MDL No.: MFCD00011232

Formula: SiO₂
Density: 1.40
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9
EINECS: 231-545-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR